LIF372154119 Life Chemicals Inc.

8-Trifluoromethyl-9H-purin-6-ylamine

  • MDL number MFCD08436659

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart