LIF372154128 Life Chemicals Inc.

2-(2,4,5-Trimethoxyphenyl)-1,3-thiazolidine

  • MDL number MFCD08138302
  • Cas Number : 896675-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart