LIF372154137 Life Chemicals Inc.

5-hydrazino-1,3-dimethylpyrido[2,3-{d}]pyrimidine-2,4(1{H},3{H})-dione

  • MDL number MFCD16652789
  • Cas Number : 112734-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart