LIF372154142 Life Chemicals Inc.

2-Chloro-4,6-dimethylpyrimidine

  • MDL number MFCD00023199
  • Cas Number : 4472-44-0
  • Molecular Weight : 142.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart