LIF372154151 Life Chemicals Inc.

8-Bromo-5-nitroquinoline

  • MDL number MFCD00509069
  • Cas Number : 139366-35-1
  • Molecular Weight : 253.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart