LIF372154190 Life Chemicals Inc.

2-methoxy-4,6-dimethylnicotinonitrile

  • MDL number MFCD00124716
  • Cas Number : 65515-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart