LIF372154197 Life Chemicals Inc.

3-(2,4-difluorophenyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD05170037
  • Cas Number : 1039053-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart