LIF372154202 Life Chemicals Inc.

3-(2-fluorophenyl)-1-methyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD16652795
  • Cas Number : 1204296-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart