LIF372154206 Life Chemicals Inc.

methyl 3-(3-bromophenyl)-1H-pyrazole-5-carboxylate

  • MDL number MFCD16652797
  • Cas Number : 1148158-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart