LIF372154212 Life Chemicals Inc.

methyl 3-(2-methoxyphenyl)-1H-pyrazole-5-carboxylate

  • MDL number MFCD03761321
  • Cas Number : 898052-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart