LIF372154283 Life Chemicals Inc.

1-(3,4-dimethylphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11920474
  • Cas Number : 1204297-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart