LIF372154305 Life Chemicals Inc.

4-(2-chlorophenyl)-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD09991881
  • Cas Number : 186195-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart