LIF372154352 Life Chemicals Inc.

[(5-ethyl-4-methyl-6-oxo-1,6-dihydropyrimidin-2-yl)thio]acetic acid

  • MDL number MFCD16631644
  • Cas Number : 347343-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart