LIF372154353 Life Chemicals Inc.

6-ethyl-2-hydrazinopyrimidin-4(3{H})-one

  • MDL number MFCD11986380
  • Cas Number : 1899-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart