LIF372154354 Life Chemicals Inc.

2-hydrazino-5,6-dimethylpyrimidin-4(3{H})-one

  • MDL number MFCD16631645
  • Cas Number : 59224-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart