LIF372154355 Life Chemicals Inc.

5-ethyl-2-hydrazino-6-methylpyrimidin-4(3H)-one

  • MDL number MFCD19201359
  • Cas Number : 65004-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart