LIF372154356 Life Chemicals Inc.

2-hydrazino-6-phenylpyrimidin-4(3{H})-one

  • MDL number MFCD00847527
  • Cas Number : 86799-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart