LIF372154357 Life Chemicals Inc.

5-ethyl-3-mercapto[1,2,4]triazolo[4,3-{a}]pyrimidin-7(8{H})-one

  • MDL number MFCD11986381
  • Cas Number : 1105189-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart