LIF372154383 Life Chemicals Inc.

5-amino-1-(4-methylphenyl)-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD11986386
  • Cas Number : 1105189-52-3
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart