LIF372154390 Life Chemicals Inc.

2-(Methoxycarbonyl)nicotinic acid

  • MDL number MFCD00111560
  • Cas Number : 24195-07-1
  • Molecular Weight : 181.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart