LIF372154414 Life Chemicals Inc.

2-Amino-7-(2-thienyl)[1,3]thiazolo[4,5-d]pyridazin-4(5H)-one

  • MDL number MFCD09743096
  • Cas Number : 941868-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart