LIF372154424 Life Chemicals Inc.

ethyl 5-amino-4-oxo-3-phenyl-3,4-dihydrothieno[3,4-d]pyridazine-1-carboxylate

  • MDL number MFCD00197075
  • Cas Number : 123542-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart