LIF372154481 Life Chemicals Inc.

1-(4-fluorophenyl)-4-methoxy-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11986416
  • Cas Number : 1169973-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart