LIF372154482 Life Chemicals Inc.

4-ethoxy-1-(4-fluorophenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11986417
  • Cas Number : 1172231-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart