LIF372154490 Life Chemicals Inc.

[5-(3-methoxyphenyl)isoxazol-3-yl]methanol

  • MDL number MFCD11868818
  • Cas Number : 1105191-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 92
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart