LIF372154491 Life Chemicals Inc.

[5-(4-methoxyphenyl)isoxazol-3-yl]methanol

  • MDL number MFCD06199341
  • Cas Number : 58492-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart