LIF372154496 Life Chemicals Inc.

[5-(thiophen-2-yl)-1,2-oxazol-3-yl]methanol

  • MDL number MFCD03086162
  • Cas Number : 194491-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart