LIF372154505 Life Chemicals Inc.

3-(chloromethyl)-5-(3-methoxyphenyl)isoxazole

  • MDL number MFCD06805615
  • Cas Number : 934063-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart