LIF372154510 Life Chemicals Inc.

methyl 5-(2-fluorophenyl)isoxazole-3-carboxylate

  • MDL number MFCD11986430
  • Cas Number : 668970-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart