LIF372154514 Life Chemicals Inc.

methyl 5-(3-methoxyphenyl)isoxazole-3-carboxylate

  • MDL number MFCD03778972
  • Cas Number : 517870-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart