LIF372154516 Life Chemicals Inc.

methyl 5-(2-methyl-2,3-dihydro-1-benzofuran-5-yl)isoxazole-3-carboxylate

  • MDL number MFCD11986433
  • Cas Number : 1105191-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart