LIF372154531 Life Chemicals Inc.

(5-phenylisoxazol-3-yl)acetonitrile

  • MDL number MFCD11986441
  • Cas Number : 35221-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart