LIF372154563 Life Chemicals Inc.

5-[(2-methyl-1H-benzimidazol-1-yl)methyl]-2-furoic acid

  • MDL number MFCD06444799
  • Cas Number : 1016763-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart