LIF372154660 Life Chemicals Inc.

5-hydroxy-2-methylisoquinolin-1(2H)-one

  • MDL number MFCD11986505
  • Cas Number : 42792-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart