LIF372154685 Life Chemicals Inc.

3-ethoxy-6-piperazin-1-ylpyridazine

  • MDL number MFCD11986518
  • Cas Number : 1105195-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart