LIF372154690 Life Chemicals Inc.

2-(2-(pyridin-2-yl)-1H-benzo[d]imidazol-1-yl)acetic acid

  • MDL number MFCD07186529
  • Cas Number : 100726-39-4
  • Molecular Weight : 253.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart