LIF372154712 Life Chemicals Inc.

ethyl 2-[(1-oxo-1,2-dihydroisoquinolin-5-yl)oxy]propanoate

  • MDL number MFCD11986536
  • Cas Number : 1014107-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 92
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart