LIF372154716 Life Chemicals Inc.

[2-(4-piperidin-1-yl-1H-pyrazolo[3,4-d]pyrimidin-1-yl)ethyl]amine

  • MDL number MFCD16652895
  • Cas Number : 1105196-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart