LIF372154726 Life Chemicals Inc.

1-(4-fluorophenyl)-4-methyl-1,6-dihydro-7H-pyrazolo[3,4-d]pyridazin-7-one

  • MDL number MFCD11986546
  • Cas Number : 1105196-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart