LIF372154730 Life Chemicals Inc.

4-methyl-1-phenyl-1,6-dihydro-7H-pyrazolo[3,4-d]pyridazin-7-one

  • MDL number MFCD11986550
  • Cas Number : 56563-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart