LIF372154746 Life Chemicals Inc.

1-(2-aminoethyl)-5-(2-fluorobenzyl)-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD11986564
  • Cas Number : 1105197-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart