LIF372154780 Life Chemicals Inc.

1-(4-fluorophenyl)-4-methyl-1H-pyrazolo[3,4-d]pyridazine-7-thiol

  • MDL number MFCD11986590
  • Cas Number : 1105198-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart