LIF372154784 Life Chemicals Inc.

ethyl 5-methyl-1-phenyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD00020741
  • Cas Number : 89193-16-8
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart