LIF372154786 Life Chemicals Inc.

3-[3,5-dimethyl-1-(4-methyl-6-oxo-1,6-dihydropyrimidin-2-yl)-1H-pyrazol-4-yl]propanoic acid

  • MDL number MFCD11986595
  • Cas Number : 1170850-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart