LIF372154787 Life Chemicals Inc.

2-hydrazino-6-isopropylpyrimidin-4(3H)-one

  • MDL number MFCD16631650
  • Cas Number : 1105198-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart