LIF372154800 Life Chemicals Inc.

2-(5-amino-3-methyl-1H-pyrazol-1-yl)-5-ethyl-6-methylpyrimidin-4(3H)-one

  • MDL number MFCD11986608
  • Cas Number : 79871-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart