LIF372154806 Life Chemicals Inc.

2-[5-amino-3-(2-thienyl)-1H-pyrazol-1-yl]-6-methylpyrimidin-4(3H)-one

  • MDL number MFCD11986615
  • Cas Number : 1171073-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart