LIF372154811 Life Chemicals Inc.

5-ethyl-6-methyl-2-sulfanyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD01677038
  • Cas Number : 39083-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart