LIF372154813 Life Chemicals Inc.

2-(5-amino-3-methyl-1H-pyrazol-1-yl)-6-phenylpyrimidin-4(3H)-one

  • MDL number MFCD11986621
  • Cas Number : 1171738-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart