LIF372154817 Life Chemicals Inc.

methyl 3-[1-(4,5-dimethyl-6-oxo-1,6-dihydropyrimidin-2-yl)-3,5-dimethyl-1H-pyrazol-4-yl]propanoate

  • MDL number MFCD11986626
  • Cas Number : 1172006-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart